Assay Details
Target Gene Details
Entrez Gene ID: | 110115 |
Gene Name: | cytochrome P450, family 11, subfamily b, polypeptide 1 |
Gene Aliases: |
AA389061, CPN1, Cyp11b, Cyp11b-1, FHI |
Location: |
Chr.15:74834892-74841643 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp11b1 | NM_001033229.3 | NP_001028401.2 | ||
AK142439.1 | BAE25066.1 | |||
AK163083.1 | BAE37185.1 | |||
AK168839.1 | BAE40662.1 | |||
BC139248.1 | ||||
BC139250.1 |
Target Gene Details
Entrez Gene ID: | 15202 |
Gene Name: | hematopoietic cell transcript 1 |
Gene Aliases: |
1700057K19Rik, HemT |
Location: |
Chr.15:74818880-74846164 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hemt1 |