Assay Details
Target Gene Details
Entrez Gene ID: | 13196 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain1 |
Gene Aliases: |
AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19 |
Location: |
Chr.15:64086840-64382919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asap1 | NM_001276461.1 | NP_001263390.1 | ||
NM_001276462.1 | NP_001263391.1 | |||
NM_001276463.1 | NP_001263392.1 | |||
NM_001276467.1 | NP_001263396.1 | |||
NM_010026.3 | NP_034156.2 | |||
AF075461.1 | AAC98349.1 | |||
AF075462.1 | AAC98350.1 | |||
AK122477.1 | BAC65759.1 | |||
AK132979.1 | ||||
AK146545.1 | BAE27250.1 | |||
AK147689.1 | ||||
BC048818.1 | AAH48818.1 | |||
BC094581.1 | AAH94581.1 | |||
CB527210.1 | ||||
CD355387.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |