Assay Details
Target Gene Details
Entrez Gene ID: | 239420 |
Gene Name: | CUB and Sushi multiple domains 3 |
Gene Aliases: |
4930500N14Rik, mKIAA1894 |
Location: |
Chr.15:47580633-48792231 on Build GRCm38 |
Assay Gene Location: | Within Exon 56 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csmd3 | NM_001081391.2 | 56 | 8973 | NP_001074860.2 |
AK122567.1 | 39 | 6156 | BAC65849.1 |