Assay Details
Target Gene Details
Entrez Gene ID: | 12181 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
AU020183, AW146150, D18861, Erb1p, Kiaa0124, mKIAA0124 |
Location: |
Chr.15:76452996-76477269 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bop1 | NM_013481.1 | NP_038509.1 | ||
AK079627.1 | BAC37708.1 | |||
AK129058.1 | BAC97868.1 | |||
AK144095.1 | ||||
AK167110.1 | BAE39258.1 | |||
AK167225.1 | BAE39350.1 | |||
AK167829.1 | BAE39851.1 | |||
AK167878.1 | ||||
AK168127.1 | BAE40096.1 | |||
BC012693.1 | AAH12693.1 | |||
U77415.1 | AAB19223.1 |
More Information
Set Membership: |
Intragenic Non-exonic |