Assay Details
Target Gene Details
Entrez Gene ID: | 66168 |
Gene Name: | glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding) |
Gene Aliases: |
1110025J15Rik, Lag, Tmbim3 |
Location: |
Chr.15:76246740-76249911 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grina | NM_023168.3 | 4 | 663 | NP_075657.1 |
AF182040.1 | 4 | 646 | AAG16897.1 | |
AK003920.1 | 4 | 672 | ||
AK185141.1 | ||||
AK187074.1 | 1 | 45 | ||
AK188174.1 | ||||
AK189399.1 | 3 | 287 | ||
AK207443.1 | ||||
AK218437.1 | ||||
BC010802.1 | 4 | 696 | AAH10802.1 | |
BC019157.1 | 4 | 684 | AAH19157.1 | |
BC037667.1 | 4 | 663 | AAH37667.2 |