Assay Details
Target Gene Details
Entrez Gene ID: | 223780 |
Gene Name: | adrenomedullin 2 |
Gene Aliases: |
Am2, IMD, Imdn |
Location: |
Chr.15:89322899-89324731 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adm2 | NM_182928.4 | 1 | 157 | NP_891558.1 |
AB121035.1 | 1 | 81 | BAD07412.1 | |
BC096428.1 | 1 | 84 | AAH96428.1 | |
CJ052708.1 | 1 | 158 | ||
DV649647.1 |
More Information
Set Membership: |
Intragenic Exonic |