Assay Details
Target Gene Details
Entrez Gene ID: | 18155 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, N23K, Npnc1, OFQ/N |
Location: |
Chr.14:65400673-65425581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnoc | NM_001205075.1 | NP_001192004.1 | ||
NM_010932.2 | NP_035062.1 | |||
AK045893.1 | BAC32523.1 | |||
BC119372.1 | ||||
BC144865.1 | ||||
D50055.1 | BAA08773.1 | |||
D50056.1 | BAA08774.1 | |||
D82866.1 | BAA11614.1 | |||
KF006993.1 | ||||
X97373.1 | CAA66042.1 |
More Information
Set Membership: |
Intragenic Non-exonic |