Assay Details
Target Gene Details
Entrez Gene ID: | 218772 |
Gene Name: | retinoic acid receptor, beta |
Gene Aliases: |
A830025K23, Hap, Nr1b2 |
Location: |
Chr.14:16430839-17082331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rarb | NM_001289760.1 | NP_001276689.1 | ||
NM_001289761.1 | NP_001276690.1 | |||
NM_001289762.1 | NP_001276691.1 | |||
NM_011243.2 | NP_035373.1 | |||
AK043726.1 | ||||
AK054178.1 | 2 | 743 | ||
AK078471.1 | ||||
AK190447.1 | ||||
BC076597.1 | AAH76597.1 | |||
S56660.1 | AAB25784.2 |
More Information
Set Membership: |
Intragenic Non-exonic |