Assay Details
Target Gene Details
Entrez Gene ID: | 69019 |
Gene Name: | signal peptidase complex subunit 1 homolog (S. cerevisiae) |
Gene Aliases: |
1810004F21Rik |
Location: |
Chr.14:30999826-31001666 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spcs1 | NM_026911.3 | NP_081187.2 | ||
AI848281.1 | ||||
AK007336.1 | BAB24969.1 | |||
AK082928.1 | BAC38696.1 | |||
AK088658.1 | 2 | 397 | ||
BC049895.1 | AAH49895.1 |
More Information
Set Membership: |
Intragenic Non-exonic |