Assay Details
Target Gene Details
Entrez Gene ID: | 19065 |
Gene Name: | neuropeptide Y receptor Y4 |
Gene Aliases: |
NYYR-D, Ppyr1, Y4 |
Location: |
Chr.14:34145646-34152419 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npy4r | NM_008919.4 | 1 | 80 | NP_032945.3 |
BC132395.1 | 1 | 80 | ||
BC145981.1 | 1 | 80 |
More Information
Set Membership: |
Intragenic Exonic |