Assay Details
Target Gene Details
Entrez Gene ID: | 628705 |
Gene Name: | PHD finger protein 11C |
Gene Aliases: |
EG628705, Gm6907, Phf11 |
Location: |
Chr.14:59380833-59440890 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf11c |
Target Gene Details
Entrez Gene ID: | 239122 |
Gene Name: | SET domain, bifurcated 2 |
Gene Aliases: |
Clld8, Gm293, KMT1F |
Location: |
Chr.14:59402011-59440877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Setdb2 | NM_001081024.1 | NP_001074493.1 | ||
AK089197.1 | 5 | 897 | BAC40789.1 | |
BC157926.1 | ||||
BC172021.1 | ||||
EU155090.1 | ||||
EU155091.1 | ||||
EU155092.1 | ||||
EU155093.1 | ||||
EU155094.1 | ||||
EU155096.1 | ||||
EU155097.1 | ||||
EU155098.1 | ||||
EU155099.1 |
More Information
Set Membership: |
Intragenic Non-exonic |