Assay Details
Target Gene Details
Entrez Gene ID: | 57811 |
Gene Name: | retinal G protein coupled receptor |
Gene Aliases: |
- |
Location: |
Chr.14:37034909-37049014 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgr | NM_001301692.1 | 5 | 650 | NP_001288621.1 |
NM_001301694.1 | 5 | 600 | NP_001288623.1 | |
NM_021340.4 | 6 | 772 | NP_067315.1 | |
AF076930.1 | 6 | 692 | AAC69836.1 | |
AK044450.1 | 6 | 724 | BAC31926.1 | |
AK044649.1 | 6 | 723 | BAC32017.1 | |
AK149272.1 | 6 | 723 | BAE28782.1 | |
BC119055.1 | 6 | 706 | ||
CK623457.1 | 5 | 531 | ||
CK626179.1 |
More Information
Set Membership: |
Intragenic Exonic |