Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gmpr2 |
Target Gene Details
Entrez Gene ID: | 28113 |
Gene Name: | Terf1 (TRF1)-interacting nuclear factor 2 |
Gene Aliases: |
AW552114, D14Wsu146e, Tin2 |
Location: |
Chr.14:55676208-55682624 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tinf2 | NM_145705.3 | NP_663751.2 | ||
AF214013.1 | AAF23504.1 | |||
AF518764.1 | AAN32986.1 | |||
AI844697.1 | ||||
AK011154.1 | ||||
AK016456.1 | ||||
AK028952.1 | ||||
AK090324.1 | ||||
AK144592.1 | BAE25954.1 | |||
AK169480.1 | 3 | 1459 | ||
AK205383.1 | ||||
AK205420.1 | ||||
AK214714.1 | ||||
BC017142.1 | ||||
BC030347.1 | AAH30347.1 |