Assay Details
Target Gene Details
Entrez Gene ID: | 71988 |
Gene Name: | establishment of cohesion 1 homolog 2 (S. cerevisiae) |
Gene Aliases: |
2410004I17Rik, D030072L07Rik |
Location: |
Chr.14:65819027-65833969 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esco2 | NM_028039.2 | 1 | 48 | NP_082315.3 |
AK050845.1 | BAC34431.1 | |||
AK144685.1 | BAE26012.1 | |||
AK166457.1 | BAE38788.1 | |||
BC033303.1 | AAH33303.2 | |||
BC071275.1 | AAH71275.1 | |||
BY029949.1 | 1 | 74 | ||
CF168369.1 |
More Information
Set Membership: |
Intragenic Exonic |