Assay Details
Target Gene Details
Entrez Gene ID: | 105511 |
Gene Name: | family with sequence similarity 170, member B |
Gene Aliases: |
4922501K12Rik, AI449756 |
Location: |
Chr.14:32833962-32836789 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam170b | NM_001164485.1 | 1 | 69 | NP_001157957.1 |
BY104921.1 | 1 | 70 |