Assay Details
Target Gene Details
Entrez Gene ID: | 211922 |
Gene Name: | DENN/MADD domain containing 6A |
Gene Aliases: |
Fam116a |
Location: |
Chr.14:26573858-26634322 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dennd6a | NM_001134465.2 | NP_001127937.1 | ||
NM_001285466.1 | NP_001272395.1 | |||
NM_001285467.1 | NP_001272396.1 | |||
NM_145969.4 | NP_666081.1 | |||
AK033036.1 | BAC28133.1 | |||
AK054197.1 | BAC35690.1 | |||
AK054295.1 | BAC35721.1 | |||
AK131163.1 | BAD21413.1 | |||
AK132219.1 | BAE21040.1 | |||
AK135953.1 | BAE22742.1 | |||
AK161908.1 | ||||
AK163396.1 | BAE37333.1 | |||
BB666315.1 | ||||
BC025603.1 | AAH25603.1 |