Assay Details
Target Gene Details
Entrez Gene ID: | 114874 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
Mir5131, PA-PLA1 |
Location: |
Chr.14:45593171-45658143 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddhd1 | NM_001039106.3 | 11 | 2380 | NP_001034195.2 |
NM_001042719.2 | 12 | 2482 | NP_001036184.1 | |
NM_001284399.1 | 13 | 2454 | NP_001271328.1 | |
NM_176845.5 | 11 | 2380 | NP_789815.3 | |
AK014975.1 | 9 | 1166 | ||
AK048690.1 | 12 | 1940 | ||
AK129425.1 | 9 | 1791 | BAC98235.1 | |
AK155294.1 | 13 | 2411 | ||
AK170271.1 | 12 | 2094 | BAE41676.1 | |
BC018552.1 | 3 | 427 | AAH18552.1 | |
BC043475.1 | 10 | 1634 | AAH43475.1 | |
CA315511.1 | 2 | 153 | ||
DQ272481.1 | 11 | 2379 | ABB76652.1 | |
DQ272482.1 | 11 | 2389 | ABB76653.1 |
More Information
Set Membership: |
Intragenic Exonic |