Assay Details
Target Gene Details
Entrez Gene ID: | 67272 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 5 |
Gene Aliases: |
1500005P16Rik, 2900052H21Rik, Cklfsf5 |
Location: |
Chr.14:54936470-54939277 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm5 | NM_026066.2 | NP_080342.1 | ||
NM_029390.1 | NP_083666.1 | |||
AK005168.1 | BAB23858.1 | |||
AK005375.1 | ||||
AK013666.1 | BAB28947.1 | |||
AY241867.1 | AAP33489.1 | |||
BC049665.1 | AAH49665.1 | |||
DQ115983.1 | AAZ20316.1 |
More Information
Set Membership: |
Intragenic Non-exonic |