Assay Details
Target Gene Details
Entrez Gene ID: | 71704 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 3 |
Gene Aliases: |
1200004I24Rik, 9830169H03Rik, C76747, Xpln |
Location: |
Chr.14:27114823-27403916 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgef3 | NM_001289686.1 | NP_001276615.1 | ||
NM_001289687.1 | NP_001276616.1 | |||
NM_001289688.1 | NP_001276617.1 | |||
NM_027871.2 | NP_082147.1 | |||
AK004600.2 | BAB23401.1 | |||
AK029872.1 | BAC26652.1 | |||
BC005517.1 | AAH05517.1 | |||
BC012262.1 | AAH12262.1 | |||
BY224523.1 | ||||
CD539300.1 |
More Information
Set Membership: |
Intragenic Non-exonic |