Assay Details
Target Gene Details
Entrez Gene ID: | 72993 |
Gene Name: | adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1 |
Gene Aliases: |
2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13 |
Location: |
Chr.14:26918986-26971205 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Appl1 | NM_145221.2 | 10 | 878 | NP_660256.1 |
AK030445.1 | 10 | 758 | ||
AK033566.1 | 10 | 849 | BAC28364.1 | |
AK045438.1 | 10 | 814 | BAC32367.1 | |
AK049307.1 | 10 | 865 | BAC33672.2 | |
AK146356.1 | 10 | 849 | BAE27108.1 | |
AY113705.1 | 10 | 843 | AAM55531.1 | |
BC019708.1 | 2 | 156 | AAH19708.1 | |
BC063751.1 | 10 | 890 | AAH63751.1 |
More Information
Set Membership: |
Intragenic Exonic |