Assay Details
Target Gene Details
Entrez Gene ID: | 210808 |
Gene Name: | laccase (multicopper oxidoreductase) domain containing 1 |
Gene Aliases: |
9030625A04Rik |
Location: |
Chr.14:77024201-77036898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lacc1 | NM_172488.2 | NP_766076.1 | ||
AK033565.1 | BAC28363.1 | |||
AK053693.1 | BAC35476.1 | |||
AK152154.1 | BAE30989.1 | |||
AK153349.1 | BAE31925.1 | |||
BC116748.1 | ||||
BC119140.1 |