Assay Details
Target Gene Details
Entrez Gene ID: | 13162 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 |
Gene Aliases: |
DAT, Dat1 |
Location: |
Chr.13:73535528-73578672 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a3 | NM_010020.3 | 9 | 1357 | NP_034150.1 |
AF109072.1 | 9 | 1280 | AAD19643.1 | |
AF109391.1 | 8 | 1205 | AAF85795.1 | |
AJ238309.1 | 8 | 1213 | CAB51926.1 | |
AK032629.1 | 9 | 1315 | BAC27959.1 | |
AK136885.1 | 9 | 1302 | BAE23154.1 | |
BC054119.1 | 9 | 1357 | AAH54119.1 |
More Information
Set Membership: |
Intragenic Exonic |