Assay Details
Target Gene Details
Entrez Gene ID: | 19210 |
Gene Name: | phosphatidylserine synthase 1 |
Gene Aliases: |
AU044268, AW539008, PSS-1, mKIAA0024 |
Location: |
Chr.13:66932830-66998401 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptdss1 | NM_008959.3 | 10 | 1802 | NP_032985.2 |
AF042731.1 | 10 | 1217 | AAB97845.1 | |
AK036990.1 | 10 | 1264 | BAC29660.1 | |
AK054101.1 | 10 | 1241 | BAC35656.1 | |
AK085857.1 | 10 | 1803 | BAC39555.1 | |
AK086136.1 | 10 | 1461 | ||
AK088048.1 | 10 | 1284 | BAC40117.1 | |
AK137681.1 | 10 | 1472 | BAE23459.1 | |
AK172873.1 | 9 | 3588 | BAD32151.1 | |
BC003260.1 | 10 | 1439 | AAH03260.1 |
More Information
Set Membership: |
Intragenic Exonic |