Assay Details
Target Gene Details
Entrez Gene ID: | 13162 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 |
Gene Aliases: |
DAT, Dat1 |
Location: |
Chr.13:73535528-73578672 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a3 | NM_010020.3 | 4 | 650 | NP_034150.1 |
AF109072.1 | 4 | 573 | AAD19643.1 | |
AF109391.1 | 3 | 498 | AAF85795.1 | |
AJ238309.1 | 3 | 506 | CAB51926.1 | |
AK032629.1 | 4 | 608 | BAC27959.1 | |
AK136885.1 | 4 | 595 | BAE23154.1 | |
BC054119.1 | 4 | 650 | AAH54119.1 |
More Information
Set Membership: |
Intragenic Exonic |