Assay Details
Target Gene Details
Entrez Gene ID: | 72391 |
Gene Name: | cyclin-dependent kinase inhibitor 3 |
Gene Aliases: |
2410006H10Rik, KAP |
Location: |
Chr.14:46760464-46771529 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdkn3 | NM_028222.1 | NP_082498.1 | ||
AK010426.1 | BAB26929.1 | |||
AK033341.1 | BAC28238.1 | |||
AK146312.1 | BAE27067.1 | |||
BC049694.1 | AAH49694.1 | |||
BY084534.1 |
More Information
Set Membership: |
Intragenic Non-exonic |