Assay Details
Target Gene Details
Entrez Gene ID: | 108912 |
Gene Name: | cell division cycle associated 2 |
Gene Aliases: |
2610311M19Rik, AI586158 |
Location: |
Chr.14:67673180-67715841 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca2 | NM_175384.4 | NP_780593.3 |