Assay Details
Target Gene Details
Entrez Gene ID: | 52882 |
Gene Name: | regulator of G-protein signalling 7 binding protein |
Gene Aliases: |
A930030I01Rik, AI842293, D13Bwg1146e, R7bp |
Location: |
Chr.13:104945904-105054930 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs7bp | NM_029879.2 | NP_084155.2 | ||
AK020910.1 | BAB32250.1 | |||
AK028323.1 | ||||
AK032138.1 | ||||
AK036240.1 | BAC29358.1 | |||
AK046733.1 | BAC32849.1 | |||
AK137417.1 | BAE23346.1 | |||
BC118957.2 | ||||
DQ104214.1 |