Assay Details
Target Gene Details
Entrez Gene ID: | 16553 |
Gene Name: | kinesin family member 13A |
Gene Aliases: |
4930505I07Rik, mKIAA4109 |
Location: |
Chr.13:46749087-46931300 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif13a | NM_010617.2 | NP_034747.2 | ||
AB037923.2 | BAB16346.1 | |||
AK029721.1 | BAC26583.1 | |||
AK077053.1 | ||||
AK220490.1 | BAD90288.1 |
More Information
Set Membership: |
Intragenic Non-exonic |