Assay Details
Target Gene Details
Entrez Gene ID: | 18260 |
Gene Name: | occludin |
Gene Aliases: |
AI503564, Ocl |
Location: |
Chr.13:100496504-100553689 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ocln | NM_008756.2 | 4 | 981 | NP_032782.1 |
AK019880.1 | 4 | 933 | BAB31900.1 | |
AK033532.1 | 4 | 914 | ||
AK042576.1 | 4 | 981 | BAC31298.1 | |
AK145798.1 | 2 | 140 | BAE26656.1 | |
BC138679.1 | 4 | 964 | ||
BC138680.1 | 4 | 964 | ||
BC145371.1 | 4 | 979 | ||
U49185.1 | 4 | 1024 | AAC52515.1 |
More Information
Set Membership: |
Intragenic Exonic |