Assay Details
Target Gene Details
Entrez Gene ID: | 18212 |
Gene Name: | neurotrophic tyrosine kinase, receptor, type 2 |
Gene Aliases: |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Location: |
Chr.13:58806569-59133970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntrk2 | NM_001025074.2 | NP_001020245.1 | ||
NM_001282961.1 | NP_001269890.1 | |||
NM_008745.3 | NP_032771.1 | |||
AB377224.1 | ||||
AB377225.1 | ||||
AK009606.1 | ||||
AK018789.1 | BAB31412.1 | |||
AK028827.1 | ||||
AK147391.1 | BAE27880.1 | |||
AK147441.1 | BAE27914.1 | |||
AK160789.1 | BAE36012.1 | |||
BC052014.1 | AAH52014.2 | |||
KC855567.1 | ||||
M33385.1 | AAA40482.1 |
More Information
Set Membership: |
Intragenic Non-exonic |