Assay Details
Target Gene Details
Entrez Gene ID: | 105348 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
2310001L02Rik, AW125446, D030064E01Rik, GP73, Golph2, PSEC0257 |
Location: |
Chr.13:59634996-59675784 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golm1 | NM_001035122.2 | 8 | 889 | NP_001030294.1 |
NM_027307.4 | 9 | 1025 | NP_081583.3 | |
AK009072.1 | 7 | 696 | ||
AK133865.1 | 8 | 891 | BAE21897.1 | |
AK167646.1 | 8 | 866 | BAE39697.1 | |
BC011152.1 | 8 | 850 | AAH11152.1 | |
BC098486.1 | 9 | 1021 | AAH98486.1 |