Assay Details
Target Gene Details
Entrez Gene ID: | 210992 |
Gene Name: | lysophosphatidylcholine acyltransferase 1 |
Gene Aliases: |
2900035H07Rik, Aytl2, BB137372, BC005662, C87117, LPCAT, rd11 |
Location: |
Chr.13:73467204-73516420 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpcat1 | NM_145376.5 | NP_663351.3 | ||
AB244717.2 | ||||
AK046079.1 | 12 | 1342 | BAC32594.1 | |
AK046507.1 | 12 | 1342 | BAC32760.1 | |
AK081865.1 | 14 | 1705 | BAC38353.1 | |
AK132379.1 | 14 | 1681 | ||
AK155286.1 | 14 | 1711 | BAE33166.1 | |
AK169190.1 | 14 | 1680 | BAE40966.1 | |
AK170723.1 | 14 | 1700 | BAE41980.1 | |
AK171582.1 | 14 | 1700 | BAE42540.1 | |
BC005662.1 | 12 | 1077 | AAH05662.1 | |
BC066809.1 | 14 | 1496 | AAH66809.1 |
More Information
Set Membership: |
Intragenic Non-exonic |