Assay Details
Target Gene Details
Entrez Gene ID: | 100039691 |
Gene Name: | long intergenic non-coding RNA, embryonic stem cells expressed 1 |
Gene Aliases: |
ENSMUSG00000078952, Gm11019, Gm2373, linc-Enc1 |
Location: |
Chr.13:97434970-97497664 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lincenc1 | NR_110430.1 | |||
NR_110431.1 | ||||
NR_110432.1 | ||||
BY736053.1 |