Assay Details
Target Gene Details
Entrez Gene ID: | 238568 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6d |
Gene Aliases: |
Gm11390, OTTMUSG00000000712, SPI3D |
Location: |
Chr.13:33661405-33671585 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinb6d | NM_001076790.2 | 5 | 771 | NP_001070258.1 |
NM_001300892.1 | 5 | 616 | NP_001287821.1 | |
AA168365.1 | 2 | 183 | ||
AK136270.1 | 5 | 773 | BAE22906.1 | |
BC147559.1 | 5 | 616 |
More Information
Set Membership: |
Intragenic Exonic |