Assay Details
Target Gene Details
Entrez Gene ID: | 66410 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
2410017I18Rik, Mterfd1 |
Location: |
Chr.13:66905456-66933072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mterf3 | NM_025547.3 | NP_079823.2 | ||
AK002299.1 | ||||
AK010525.1 | BAB27006.1 | |||
AK011934.1 | BAB27924.3 | |||
AK034645.1 | ||||
BC025173.1 | AAH25173.1 | |||
BC085282.1 | AAH85282.1 |