Assay Details
Target Gene Details
Entrez Gene ID: | 218236 |
Gene Name: | family with sequence similarity 120, member A |
Gene Aliases: |
C9orf10, Ossa |
Location: |
Chr.13:48879216-48967877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam120a | NM_001033268.2 | NP_001028440.2 | ||
AK172909.1 | BAD32187.1 | |||
BC010304.1 | AAH10304.1 | |||
BC042582.1 | ||||
BC158134.1 |