Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc6 | NM_025695.4 | 9 | 1209 | NP_079971.2 |
AJ310552.1 | 8 | 801 | CAC39250.1 | |
AK009009.1 | 9 | 943 | BAB26022.3 | |
AK011982.1 | 9 | 929 | BAB27956.1 | |
AK019823.1 | 5 | 336 | ||
AK050341.1 | 9 | 903 | BAC34200.2 | |
AK088002.1 | 9 | 967 | BAC40087.2 | |
AK135873.1 | 9 | 947 | BAE22709.1 | |
AK148478.1 | 9 | 974 | BAE28576.1 | |
AK220488.1 | 8 | 884 | BAD90287.1 | |
BC048790.1 | 9 | 966 | AAH48790.1 | |
BC090630.1 | 9 | 1209 | AAH90630.1 | |
BC099955.1 | 9 | 896 | AAH99955.1 |
More Information
Set Membership: |
Intragenic Exonic |