Assay Details
Target Gene Details
Entrez Gene ID: | 328110 |
Gene Name: | PRP39 pre-mRNA processing factor 39 homolog (yeast) |
Gene Aliases: |
Srcs1 |
Location: |
Chr.12:65036309-65063386 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prpf39 | NM_177806.3 | NP_808474.2 | ||
AK154170.1 | BAE32419.1 | |||
AK168462.1 | ||||
BC014689.1 | ||||
BC029153.1 | AAH29153.1 |
More Information
Set Membership: |
Intragenic Non-exonic |