Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc6 | NM_025695.4 | NP_079971.2 | ||
AJ310552.1 | CAC39250.1 | |||
AK014442.1 | BAB29353.2 | |||
AK220488.1 | BAD90287.1 | |||
BC026429.1 | AAH26429.1 | |||
BC090630.1 | AAH90630.1 |
More Information
Set Membership: |
Intragenic Non-exonic |