Assay Details
Target Gene Details
Entrez Gene ID: | 69675 |
Gene Name: | peroxidasin homolog (Drosophila) |
Gene Aliases: |
2310075M15Rik, C85409, E330004E07, VPO1, mKIAA0230 |
Location: |
Chr.12:29937877-30017658 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pxdn | NM_181395.2 | 7 | 734 | NP_852060.2 |
AK054255.1 | 6 | 649 | BAC35705.1 | |
AK122223.1 | 7 | 504 | BAC65505.1 | |
AK142872.1 | 7 | 734 | BAE25216.1 | |
AK163597.1 | 7 | 721 | BAE37412.1 | |
BC150789.1 | 7 | 729 |
More Information
Set Membership: |
Intragenic Exonic |