Assay Details
Target Gene Details
Entrez Gene ID: | 268591 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 5 |
Gene Aliases: |
4933415L04, PAI-3, Pci |
Location: |
Chr.12:104099313-104106138 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina5 | NM_172953.3 | NP_766541.2 | ||
AK077160.1 | BAC36651.1 | |||
AK087714.1 | BAC39979.1 | |||
BC090981.1 | AAH90981.1 |
More Information
Set Membership: |
Intragenic Non-exonic |