Assay Details
Target Gene Details
Entrez Gene ID: | 11624 |
Gene Name: | aryl-hydrocarbon receptor repressor |
Gene Aliases: |
mKIAA1234 |
Location: |
Chr.13:74211117-74292330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahrr | NM_009644.2 | NP_033774.1 | ||
AB015140.1 | BAA37132.1 | |||
AK154152.1 | BAE32410.1 | |||
AK154761.1 | BAE32810.1 | |||
AK155711.1 | BAE33396.1 | |||
BC118534.1 |
More Information
Set Membership: |
Intragenic Non-exonic |