Assay Details
Target Gene Details
Entrez Gene ID: | 11624 |
Gene Name: | aryl-hydrocarbon receptor repressor |
Gene Aliases: |
mKIAA1234 |
Location: |
Chr.13:74211117-74292330 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahrr | AK154761.1 | BAE32810.1 | ||
AK155711.1 | BAE33396.1 |
More Information
Set Membership: |
Intragenic Non-exonic |