Assay Details
Target Gene Details
Entrez Gene ID: | 19276 |
Gene Name: | protein tyrosine phosphatase, receptor type, N polypeptide 2 |
Gene Aliases: |
4930425H11Rik, IA2beta, PTP-NP, Phol, mKIAA0387, phogrin |
Location: |
Chr.12:116485692-117278167 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprn2 | NM_011215.2 | NP_035345.2 | ||
AK081267.1 | ||||
AK138655.1 | BAE23734.1 | |||
AK220255.1 | BAD90180.1 | |||
BC133678.1 | ||||
U57345.1 | AAB06945.1 | |||
U82439.1 | AAB39996.1 |
More Information
Set Membership: |
Intragenic Non-exonic |