Assay Details
Target Gene Details
Entrez Gene ID: | 11992 |
Gene Name: | AU RNA binding protein/enoyl-coenzyme A hydratase |
Gene Aliases: |
C77140, W91705 |
Location: |
Chr.13:52835102-52929706 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Auh | NM_016709.2 | 7 | 668 | NP_057918.2 |
AK003219.1 | 7 | 648 | ||
BC026525.1 | 7 | 606 | AAH26525.1 | |
BC049597.1 | 7 | 652 | AAH49597.1 |
More Information
Set Membership: |
Intragenic Exonic |