Assay Details
Target Gene Details
Entrez Gene ID: | 77219 |
Gene Name: | prostaglandin reductase 2 |
Gene Aliases: |
1810016I24Rik, 9130222H03Rik, 9630002F03Rik, AI838763, B830026H24Rik, PGR-2, PRG-2, Zadh1 |
Location: |
Chr.12:84285237-84315832 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptgr2 | NM_001252625.1 | 3 | 380 | NP_001239554.1 |
NM_001252626.1 | 3 | 323 | NP_001239555.1 | |
NM_029880.3 | 3 | 323 | NP_084156.2 | |
AK007522.1 | 3 | 206 | ||
AK020666.1 | 3 | 252 | ||
AK021033.1 | 3 | 215 | BAB32284.1 | |
AK036168.1 | 3 | 245 | BAC29329.1 | |
AK040396.1 | 3 | 255 | ||
AK145232.1 | 3 | 271 | BAE26315.1 | |
AK159187.1 | 3 | 219 | BAE34884.1 | |
AK159932.1 | 3 | 220 | BAE35493.1 | |
AK168895.1 | 3 | 214 | BAE40712.1 | |
BC021466.1 | 3 | 269 | AAH21466.1 | |
CJ050177.1 | 3 | 324 | ||
CJ137771.1 | 3 | 388 |
More Information
Set Membership: |
Intragenic Exonic |