Assay Details
Target Gene Details
Entrez Gene ID: | 73389 |
Gene Name: | high mobility group box transcription factor 1 |
Gene Aliases: |
1700058O05Rik, BE629963, C330012F01Rik, C86454 |
Location: |
Chr.12:31926455-31950535 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hbp1 | NM_153198.2 | NP_694878.2 | ||
NM_177993.3 | NP_818774.2 | |||
AK028674.1 | BAC26060.1 | |||
AK028949.1 | ||||
AK053005.1 | ||||
AK132503.1 | ||||
AK133127.1 | BAE21520.1 | |||
AK139211.1 | BAE23919.1 | |||
AK142187.1 | BAE24968.1 | |||
BC026853.1 | AAH26853.1 |