Assay Details
Target Gene Details
Entrez Gene ID: | 67217 |
Gene Name: | L-3-hydroxyproline dehydratase (trans-) |
Gene Aliases: |
2810055F11Rik |
Location: |
Chr.12:72071660-72085433 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L3hypdh |