Assay Details
Target Gene Details
Entrez Gene ID: | 75483 |
Gene Name: | cytochrome c oxidase subunit VIIIc |
Gene Aliases: |
1700007F21Rik, COX8-3, COXVIII-3 |
Location: |
Chr.12:102899306-102900534 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox8c | NM_001039049.1 | 1 | 50 | NP_001034138.1 |
AK005708.1 | 1 | 41 | BAC25121.1 | |
BC145768.1 | ||||
BC145770.1 | ||||
CF106409.1 | 1 | 50 |
More Information
Set Membership: |
Intragenic Exonic |