Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa1 | NM_010881.2 | 20 | 4702 | NP_035011.1 |
AK035922.1 | 5 | 1026 | BAC29244.1 | |
BC068177.1 | 17 | 4010 | AAH68177.1 | |
BC080866.1 | AAH80866.1 | |||
U56920.1 | 19 | 4260 | AAB01228.1 | |
U64606.1 | 17 | 4005 | AAB06177.1 | |
U64828.1 | 20 | 4615 | AAB38841.1 |
More Information
Set Membership: |
Intragenic Exonic |